F4/80 Monoclonal Antibody (BM8), PE-eFluor™ 610, eBioscience™ - FAQs
F4/80 Monoclonal Antibody (BM8), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for F4/80 Monoclonal Antibody (BM8), PE-eFluor™ 610, eBioscience™ - FAQs (61-4801-80, 61-4801-82)