EOMES Monoclonal Antibody (Dan11mag), PE-eFluor™ 610, eBioscience™ - FAQs
EOMES Monoclonal Antibody (Dan11mag), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (Dan11mag), PE-eFluor™ 610, eBioscience™ - FAQs (61-4875-82, 61-4875-80)