EOMES Monoclonal Antibody (WD1928), PE-eFluor™ 610, eBioscience™ - FAQs
EOMES Monoclonal Antibody (WD1928), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for EOMES Monoclonal Antibody (WD1928), PE-eFluor™ 610, eBioscience™ - FAQs (61-4877-41, 61-4877-42)