Armenian Hamster IgG Isotype Control (eBio299Arm), PE-eFluor™ 610, eBioscience™ - FAQs
Armenian Hamster IgG Isotype Control (eBio299Arm), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Armenian Hamster IgG Isotype Control (eBio299Arm), PE-eFluor™ 610, eBioscience™ - FAQs (61-4888-80, 61-4888-82)