Syrian Hamster IgG Isotype Control, PE-eFluor™ 610, eBioscience™ - FAQs
Syrian Hamster IgG Isotype Control, PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Syrian Hamster IgG Isotype Control, PE-eFluor™ 610, eBioscience™ - FAQs (61-4914-82, 61-4914-80)