Ki-67 Monoclonal Antibody (SolA15), PE-eFluor™ 610, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (SolA15), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (SolA15), PE-eFluor™ 610, eBioscience™ - FAQs (61-5698-80, 61-5698-82)