Ki-67 Monoclonal Antibody (20Raj1), PE-eFluor™ 610, eBioscience™ - FAQs
Ki-67 Monoclonal Antibody (20Raj1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Ki-67 Monoclonal Antibody (20Raj1), PE-eFluor™ 610, eBioscience™ - FAQs (61-5699-42, 61-5699-41)