FOXP3 Monoclonal Antibody (FJK-16s), PE-eFluor™ 610, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (FJK-16s), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (FJK-16s), PE-eFluor™ 610, eBioscience™ - FAQs (61-5773-82, 61-5773-80)