Mouse IgM Monoclonal Antibody (II/41), PE-eFluor™ 610, eBioscience™ - FAQs
Mouse IgM Monoclonal Antibody (II/41), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Mouse IgM Monoclonal Antibody (II/41), PE-eFluor™ 610, eBioscience™ - FAQs (61-5790-80, 61-5790-82)