CD357 (AITR/GITR) Monoclonal Antibody (eBioAITR), PE-eFluor™ 610, eBioscience™ - FAQs
CD357 (AITR/GITR) Monoclonal Antibody (eBioAITR), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD357 (AITR/GITR) Monoclonal Antibody (eBioAITR), PE-eFluor™ 610, eBioscience™ - FAQs (61-5875-41, 61-5875-42)