CD314 (NKG2D) Monoclonal Antibody (CX5), PE-eFluor™ 610, eBioscience™ - FAQs
CD314 (NKG2D) Monoclonal Antibody (CX5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD314 (NKG2D) Monoclonal Antibody (CX5), PE-eFluor™ 610, eBioscience™ - FAQs (61-5882-80, 61-5882-82)