KLRG1 Monoclonal Antibody (2F1), PE-eFluor™ 610, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (2F1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (2F1), PE-eFluor™ 610, eBioscience™ - FAQs (61-5893-80, 61-5893-82)