FceR1 alpha Monoclonal Antibody (MAR-1), PE-eFluor™ 610, eBioscience™ - FAQs
FceR1 alpha Monoclonal Antibody (MAR-1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for FceR1 alpha Monoclonal Antibody (MAR-1), PE-eFluor™ 610, eBioscience™ - FAQs (61-5898-82, 61-5898-80)