iNOS Monoclonal Antibody (CXNFT), PE-eFluor™ 610, eBioscience™ - FAQs
iNOS Monoclonal Antibody (CXNFT), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for iNOS Monoclonal Antibody (CXNFT), PE-eFluor™ 610, eBioscience™ - FAQs (61-5920-80, 61-5920-82)