TER-119 Monoclonal Antibody (TER-119), PE-eFluor™ 610, eBioscience™ - FAQs
TER-119 Monoclonal Antibody (TER-119), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for TER-119 Monoclonal Antibody (TER-119), PE-eFluor™ 610, eBioscience™ - FAQs (61-5921-82, 61-5921-80)