Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE-eFluor™ 610, eBioscience™ - FAQs
Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Ly-6G/Ly-6C Monoclonal Antibody (RB6-8C5), PE-eFluor™ 610, eBioscience™ - FAQs (61-5931-80, 61-5931-82)