NK1.1 Monoclonal Antibody (PK136), PE-eFluor™ 610, eBioscience™ - FAQs
NK1.1 Monoclonal Antibody (PK136), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for NK1.1 Monoclonal Antibody (PK136), PE-eFluor™ 610, eBioscience™ - FAQs (61-5941-82, 61-5941-80)