TCR beta Monoclonal Antibody (H57-597), PE-eFluor™ 610, eBioscience™ - FAQs
TCR beta Monoclonal Antibody (H57-597), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for TCR beta Monoclonal Antibody (H57-597), PE-eFluor™ 610, eBioscience™ - FAQs (61-5961-82, 61-5961-80)