IL-2 Monoclonal Antibody (MQ1-17H12), PE-eFluor™ 610, eBioscience™ - FAQs
IL-2 Monoclonal Antibody (MQ1-17H12), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IL-2 Monoclonal Antibody (MQ1-17H12), PE-eFluor™ 610, eBioscience™ - FAQs (61-7029-42)