IL-13 Monoclonal Antibody (eBio13A), PE-eFluor™ 610, eBioscience™ - FAQs
IL-13 Monoclonal Antibody (eBio13A), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IL-13 Monoclonal Antibody (eBio13A), PE-eFluor™ 610, eBioscience™ - FAQs (61-7133-82, 61-7133-80)