IL-17A Monoclonal Antibody (eBio17B7), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio17B7), PE-eFluor™ 610, eBioscience™ - FAQs (61-7177-80, 61-7177-82)