IL-17A Monoclonal Antibody (eBio64DEC17), PE-eFluor™ 610, eBioscience™ - FAQs
IL-17A Monoclonal Antibody (eBio64DEC17), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IL-17A Monoclonal Antibody (eBio64DEC17), PE-eFluor™ 610, eBioscience™ - FAQs (61-7179-42, 61-7179-41)