CD185 (CXCR5) Monoclonal Antibody (SPRCL5), PE-eFluor™ 610, eBioscience™ - FAQs
CD185 (CXCR5) Monoclonal Antibody (SPRCL5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD185 (CXCR5) Monoclonal Antibody (SPRCL5), PE-eFluor™ 610, eBioscience™ - FAQs (61-7185-82, 61-7185-80)