IFN gamma Monoclonal Antibody (XMG1.2), PE-eFluor™ 610, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (XMG1.2), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (XMG1.2), PE-eFluor™ 610, eBioscience™ - FAQs (61-7311-80, 61-7311-82)