IFN gamma Monoclonal Antibody (4S.B3), PE-eFluor™ 610, eBioscience™ - FAQs
IFN gamma Monoclonal Antibody (4S.B3), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IFN gamma Monoclonal Antibody (4S.B3), PE-eFluor™ 610, eBioscience™ - FAQs (61-7319-41, 61-7319-42)