TNF alpha Monoclonal Antibody (MP6-XT22), PE-eFluor™ 610, eBioscience™ - FAQs
TNF alpha Monoclonal Antibody (MP6-XT22), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for TNF alpha Monoclonal Antibody (MP6-XT22), PE-eFluor™ 610, eBioscience™ - FAQs (61-7321-80, 61-7321-82)