Granzyme B Monoclonal Antibody (NGZB), PE-eFluor™ 610, eBioscience™ - FAQs
Granzyme B Monoclonal Antibody (NGZB), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Granzyme B Monoclonal Antibody (NGZB), PE-eFluor™ 610, eBioscience™ - FAQs (61-8898-80, 61-8898-82)