KLRG1 Monoclonal Antibody (13F12F2), PE-eFluor™ 610, eBioscience™ - FAQs
KLRG1 Monoclonal Antibody (13F12F2), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for KLRG1 Monoclonal Antibody (13F12F2), PE-eFluor™ 610, eBioscience™ - FAQs (61-9488-42, 61-9488-41)