TIGIT Monoclonal Antibody (MBSA43), PE-eFluor™ 610, eBioscience™ - FAQs
TIGIT Monoclonal Antibody (MBSA43), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for TIGIT Monoclonal Antibody (MBSA43), PE-eFluor™ 610, eBioscience™ - FAQs (61-9500-41, 61-9500-42)