IRF4 Monoclonal Antibody (3E4), PE-eFluor™ 610, eBioscience™ - FAQs
IRF4 Monoclonal Antibody (3E4), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for IRF4 Monoclonal Antibody (3E4), PE-eFluor™ 610, eBioscience™ - FAQs (61-9858-82, 61-9858-80)