HELIOS Monoclonal Antibody (22F6), PE-eFluor™ 610, eBioscience™ - FAQs
HELIOS Monoclonal Antibody (22F6), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for HELIOS Monoclonal Antibody (22F6), PE-eFluor™ 610, eBioscience™ - FAQs (61-9883-42, 61-9883-41)