HLA-DR Monoclonal Antibody (LN3), PE-eFluor™ 610, eBioscience™ - FAQs
HLA-DR Monoclonal Antibody (LN3), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for HLA-DR Monoclonal Antibody (LN3), PE-eFluor™ 610, eBioscience™ - FAQs (61-9956-41, 61-9956-42)