Gata-3 Monoclonal Antibody (TWAJ), PE-eFluor™ 610, eBioscience™ - FAQs
Gata-3 Monoclonal Antibody (TWAJ), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Gata-3 Monoclonal Antibody (TWAJ), PE-eFluor™ 610, eBioscience™ - FAQs (61-9966-42, 61-9966-41)