CD279 (PD-1) Monoclonal Antibody (J43), PE-eFluor™ 610, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (J43), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (J43), PE-eFluor™ 610, eBioscience™ - FAQs (61-9985-80, 61-9985-82)