CD184 (CXCR4) Monoclonal Antibody (2B11), PE-eFluor™ 610, eBioscience™ - FAQs
CD184 (CXCR4) Monoclonal Antibody (2B11), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD184 (CXCR4) Monoclonal Antibody (2B11), PE-eFluor™ 610, eBioscience™ - FAQs (61-9991-82, 61-9991-80)