6100-01 - FAQs
6100-01 - FAQs
View additional product information for 6100-01 - FAQs (6100-01)
1 product FAQs found
I am using the ABI PRISM 6100 Nucleic Acid PrepStation. Does Thermo Fisher Scientific have recommendations for the input mass of my sample type for RNA purification?
Please refer to the protocol, Tissue RNA Isolation: Isolation of Total RNA from Plant and Animal Tissue and the Total RNA Isolation Quick Reference Card.