Cyclohexane, for HPLC, Thermo Scientific Chemicals
Cyclohexane, for HPLC, Thermo Scientific Chemicals
Cyclohexane, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cyclohexane, for HPLC, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantity
6100400404 L
Catalog number 610040040
Price (USD)
294.65
Online Exclusive
327.00
Save 32.35 (10%)
Each
Quantity:
4 L
Request bulk or custom format
Price (USD)
294.65
Online Exclusive
327.00
Save 32.35 (10%)
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.0 %
Color scale=<10 APHA
Identificationpasses test
Refractive index1.4235 to 1.4255 (25°C, 589 nm)
Residue after evaporation=<10 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only