Toluene, for HPLC, Thermo Scientific Chemicals
Toluene, for HPLC, Thermo Scientific Chemicals
Toluene, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Toluene, for HPLC, Thermo Scientific Chemicals

 
Have Questions?
Catalog NumberQuantityPackaging
6101100404 LEcoSafPak™
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 610110040
Price (USD)
-
Quantity:
4 L
Packaging:
EcoSafPak™
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 286 nm A: =<1
Color scale=<10 APHA
Refractive index1.4930 to 1.4950 (25°C, 589 nm)
Water=<0.02 %
GC>=99.8 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only