N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™
N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™
Thermo Scientific Chemicals

N,N-Dimethylformamide, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™

CAS: 68-12-2 | C3H7NO | 73.10 g/mol
Have Questions?
Catalog NumberQuantity
610941000100 mL
Catalog number 610941000
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Chemical Identifiers
CAS68-12-2
IUPAC NameN,N-dimethylformamide
Molecular FormulaC3H7NO
InChI KeyZMXDDKWLCZADIW-UHFFFAOYSA-N
SMILESCN(C)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless
Appearance (Form)Liquid
Identificationpasses test
Water=<50 ppm
Assay>=99.8 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only