This product is animal origin and can only be shipped within the United States
Product Image
Thermo Scientific Chemicals

LB Broth, Miller

 
Have Questions?
Catalog NumberQuantity
611875000500 g
Catalog number 611875000
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Specifications
ColorLight Beige
DescriptionpH at 25°C is 6.8-7.2 (2.5% solution)
Media TypeLB Broth
Packaging TypePouch
Quantity500 g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Contents & Storage
4°C