What is the composition of LB Broth, Lennox (Cat. No. 612725000)?
LB Broth, Lennox (Cat. No. 612725000) is composed of 10.0 g/L casein digest peptone, 5.0 g/L sodium chloride, and 5.0 g/L yeast extract.
Search
View additional product information for LB Broth, Lennox - FAQs (612725000)
2 product FAQs found