Nalgene™ HDPE Quick Disconnects
Thermo Scientific™
Nalgene™ HDPE Quick Disconnects
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ Quick Disconnects.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
6150-00203/8 to 7/16 in. I.D.
6150-00101/4 to 5/16 in. I.D.
6150-0010PK1/4 to 5/16 in. I.D.
6150-0020PK3/8 to 7/16 in. I.D.
Catalog number 6150-0020
Price (USD)
568.00
Case of 72
For Use With:
3/8 to 7/16 in. I.D.
Price (USD)
568.00
Case of 72
Create rapid, smooth connections between two fluid transfer paths with Thermo Scientific™ Nalgene™ HDPE Quick Disconnects, which allow tubulations to slide together snugly with a twist.
Two tapered tubulations fit tightly together with male/female connection, allowing for rapid joining or separation of apparatus lines.
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With3/8 to 7/16 in. I.D.
Packaging Quantity12/Pk., 72/Cs.
MaterialHDPE
Product TypeQuick Disconnect
Unit SizeCase of 72