Nalgene™ Tubing T-Type Connectors
Thermo Scientific™
Nalgene™ Tubing T-Type Connectors
Maximize liquid flow with non-tapered hose barbed T-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
6151-0125Tubing I.D. 1/8 in.
6151-0125PKTubing I.D. 1/8 in.
6151-0187Tubing I.D. 3/16 in.
6151-0187PKTubing I.D. 3/16 in.
6151-0250Tubing I.D. 1/4 in.
6151-0250PKTubing I.D. 1/4 in.
6151-0312Tubing I.D. 5/16 in.
6151-0312PKTubing I.D. 5/16 in.
6151-0375Tubing I.D. 3/8 in.
6151-0375PKTubing I.D. 3/8 in.
6151-0500Tubing I.D. 1/2 in.
6151-0500PKTubing I.D. 1/2 in.
Catalog number 6151-0125
Price (USD)
243.00
Case of 72
For Use With:
Tubing I.D. 1/8 in.
Price (USD)
243.00
Case of 72
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing T-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionT-Type Connector
For Use WithTubing I.D. 1/8 in.
Packaging Quantity12/Pk., 72/Cs.
AutoclavableAutoclavable
ColorClear
Diameter (English) Outer1/8 in.
Diameter (Metric) Outer3.175 mm
MaterialPolypropylene
Overall Length (English)0.81 in.
Overall Length (Metric)20.57 mm
Product TypeTubing Connector
Unit SizeCase of 72