Nalgene™ Tubing T-Type Connectors
Thermo Scientific™
Nalgene™ Tubing T-Type Connectors
Maximize liquid flow with non-tapered hose barbed T-Type Connectors, molded of polypropylene resin, free of animal derived components.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With
6151-0500Tubing I.D. 1/2 in.
6151-0125Tubing I.D. 1/8 in.
6151-0125PKTubing I.D. 1/8 in.
6151-0187Tubing I.D. 3/16 in.
6151-0187PKTubing I.D. 3/16 in.
6151-0250Tubing I.D. 1/4 in.
6151-0250PKTubing I.D. 1/4 in.
6151-0312Tubing I.D. 5/16 in.
6151-0312PKTubing I.D. 5/16 in.
6151-0375Tubing I.D. 3/8 in.
6151-0375PKTubing I.D. 3/8 in.
6151-0500PKTubing I.D. 1/2 in.
Catalog number 6151-0500
Price (USD)
259.00
Case of 48
For Use With:
Tubing I.D. 1/2 in.
Price (USD)
259.00
Case of 48
Maximize liquid flow with non-tapered hose barbed Thermo Scientific™ Nalgene™ Tubing T-Type Connectors, molded of polypropylene resin, free of animal derived components. Products are double-bagged to maintain cleanliness.
  • Autoclavable
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use WithTubing I.D. 1/2 in.
Packaging Quantity12/Pk., 72/Cs.
AutoclavableAutoclavable
Diameter (English) Outer1/2 in.
Diameter (Metric) Outer12.7 mm
MaterialPolypropylene
Overall Length (English)3.04 in.
Overall Length (Metric)77.21 mm
Product TypeTubing Connector
Unit SizeCase of 48