Search
Search
| Catalog Number | Product Type | Packaging |
|---|---|---|
| 6168-30 | External 8-Port USB Serial Box | Ea. |
| 6420-30 | Stylus | 4/Pk. |
| 6422-30 | Stylus Holder | Ea. |
| 6444-30 | Absorbent Sheet | Ea. |
| 6424-30 | Bottle Carrying Rack | Ea. |
| 6431-30 | Extension Cable | Ea. |
| 6430-30 | Remote Alarm | Ea. |
| 6434-30 | Remote Alarm Speakers | Ea. |
| 6020-30 | VersaTREK 240 System Cart | Ea. |
| 6432-30 | Internal Speaker | Ea. |
| 6440-30 | Thermometer | Ea. |
| 6705-30 | Thermometer Mount | Ea. |
| 6452-30 | Drawer Number Labels | Ea. |
| 6031-30Y | Drawer-Right | Ea. |
| 6030-30Y | Drawer-Left | Ea. |
| 6410-30 | VersaTREK/ESP User Manual (CD) | Ea. |
| 6400-30 | VersaTREK User Manual (CD) | Ea. |
| 6150-30 | HP Laserjet Printer | Ea. |
| 6152-30 | 6' USB Serial Printer Cable | Ea. |
| 6852-30Y | Power Conditioner, 360VA | Ea. |
| 6159-30 | Ethernet Switch | Ea. |
| 6170-30 | AT Adapter Cable | Ea. |
| 6172-30 | Plenum Patch Cable | Ea., 25 ft. |
| 6171-30 | Snagless Patch Cable | Ea. |
| 6174-30 | Plenum Patch Cable | Ea., 50 ft. |
| 6175-30 | Plenum Patch Cable | Ea., 100 ft. |
| 6854-30Y | Power Conditioner, 1440/1500VA | Ea. |
| 6320-30 | Seismic Restrainer Bracket 528 | Ea. |
| 6040-30 | Myco Bottle Adapters | 25/bx. |
| 6044-30 | Disabled Location Caps | Ea. |
| 6118-30-1 | Hard Drive | Ea. |
| 6119-30-1 | Hard Drive Caddy | Ea. |