Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Nalgene™ Polypropylene Stirring Rod
Thermo Scientific™

Nalgene™ Polypropylene Stirring Rod

Combine functions of a stirring rod and rubber policeman with these polypropylene stirring rods, featuring a flat paddle at one end and a triangular paddle at the other end.
Have Questions?
Catalog NumberLength (Metric)
6169-0010248 mm
Catalog number 6169-0010
Price (USD)
378.88
Online exclusive
468.18
Save 89.30
Case of 72
Length (Metric):
248 mm
Price (USD)
378.88
Online exclusive
468.18
Save 89.30
Case of 72

Combine the functions of a stirring rod and rubber policeman with Thermo Scientific™ Nalgene™ Polypropylene Stirring Rods, featuring a flat paddle at one end and a triangular paddle at the other end. These autoclavable stirring rods are chemically compatible with a wide range of lab reagents.

  • Flat paddle, 5/8 x 5/8″ (1.6 x 1.6cm) at one end; triangular paddle at the other end
  • Overall length is 9.75″ (24.8cm), makes it a convenient and conventional length for a wide variety of lab uses
  • Usable over a temperature range of 0°C to +135°C making it suitable for use in heated applications

Specifications
AutoclavableAutoclavable
ColorWhite
DescriptionStirring Rod with flat paddle and triangular paddle ends
Length (English)9¾ in.
Length (Metric)248 mm
MaterialPolypropylene
TypeStirring Rod
Unit SizeCase of 72