DMEM, high glucose, GlutaMAX™ Supplement, 500 mL - FAQs
DMEM, high glucose, GlutaMAX™ Supplement, 500 mL - FAQs
View additional product information for DMEM, high glucose, GlutaMAX™ Supplement - FAQs (61965026)
4 product FAQs found
Do both the DMEM, high glucose, GlutaMAX Supplements, Cat. No. 61965026 and 10566016, have the same formulation?
Yes, both the DMEM, high glucose, GlutaMAX Supplements, Cat. No. 61965026 and 10566016, have the same formulation.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?
Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will the media formulations containing GlutaMAX supplement change with respect to L-glutamine content?
In all media containing GlutaMAX supplement dipeptides as a substitute for L-glutamine, concentration is equimolar with the L-glutamine in the original formulation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.