DMEM, high glucose, GlutaMAX™ supplement, 10 x 500 mL - FAQs
DMEM, high glucose, GlutaMAX™ supplement, 10 x 500 mL - FAQs
View additional product information for DMEM, high glucose, GlutaMAX™ Supplement - FAQs (61965059)
2 product FAQs found
Do both the DMEM, high glucose, GlutaMAX Supplements, Cat. No. 61965026 and 10566016, have the same formulation?
Yes, both the DMEM, high glucose, GlutaMAX Supplements, Cat. No. 61965026 and 10566016, have the same formulation.
Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.
What is the manganese concentration in DMEM? Do you offer manganese-free DMEM?
Manganese is not present in the formulation of our catalog DMEM media products.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.