DRAQ5™ Fluorescent Probe Solution (5 mM), 1 mL - FAQs
DRAQ5™ Fluorescent Probe Solution (5 mM), 1 mL - FAQs
View additional product information for DRAQ5™ Fluorescent Probe Solution (5 mM) - FAQs (62251, 62252, 62254)
1 product FAQs found
Does DRAQ5 Fluorescent Probe Solution contain DMSO?
The aqueous solution that DRAQ5 Fluorescent Probe Solution (Cat. No. 62254) is manufactured in is proprietary.