Pierce™ G2 Fast Blotter Cassette - FAQs
Pierce™ G2 Fast Blotter Cassette - FAQs
View additional product information for Pierce™ G2 Fast Blotter Cassette - FAQs (62289)
1 product FAQs found
I have the Thermo Scientific G2 Fast Blotter and want to add the fast stain functionality. What should I get?
You can purchase the Thermo Scientific G2 Fast Blotter - Power Stainer Upgrade Kit (Cat. No. 62288). This kit contains the Thermo Scientific Power Stain Cassette and a USB Flash Drive loaded with software that will update the Thermo Scientific G2 Fast Blotter Control Unit software and add the staining functions. We also offer Thermo Scientific Power Staining Kits (Cat. Nos. 22839, 22840).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.